ENA390347505 Enamine

4-(aminomethyl)-N,N-dimethylpyrimidin-2-amine

  • MDL number MFCD10697044
  • Cas Number : 944902-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126468
Availability details for
Quantity
  • Availability
  • Add to Cart