ENA390347511 Enamine

6-chloro-2,3,4,9-tetrahydro-1H-carbazol-3-ol

  • MDL number MFCD21218875
  • Cas Number : 32202-79-2
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126475
Availability details for
Quantity
  • Availability
  • Add to Cart