ENA390347512 Enamine

6-(2,2-difluoroethoxy)pyridine-2-carboxylic acid

  • MDL number MFCD23701784
  • Cas Number : 1423754-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120465

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120465
Availability details for
Quantity
  • Availability
  • Add to Cart