ENA390347514 Enamine

2-(N,1-diphenylformamido)acetic acid

  • MDL number MFCD01861478
  • Cas Number : 119656-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125113
Availability details for
Quantity
  • Availability
  • Add to Cart