ENA390347519 Enamine

3-methoxy-5-(methylsulfanyl)-1,2-thiazole-4-carbonitrile

  • MDL number MFCD03017773
  • Cas Number : 157139-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126487
Availability details for
Quantity
  • Availability
  • Add to Cart