ENA390347521 Enamine

4-methoxy-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD00784582
  • Cas Number : 169945-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120487
Availability details for
Quantity
  • Availability
  • Add to Cart