ENA390347522 Enamine

5-chloro-7-methyl-1-benzofuran-2-carbonitrile

  • MDL number MFCD16743023
  • Cas Number : 1250393-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126498

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126498
Availability details for
Quantity
  • Availability
  • Add to Cart