ENA390347524 Enamine

4-{1H-pyrrolo[2,3-b]pyridin-4-yl}-1,2,3,6-tetrahydropyridine dihydrochloride

  • MDL number MFCD23701830
  • Cas Number : 1428030-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125118
Availability details for
Quantity
  • Availability
  • Add to Cart