ENA390347525 Enamine

2-amino-2,3,3-trimethylbutanenitrile

  • MDL number MFCD09735463
  • Cas Number : 52161-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126534
Availability details for
Quantity
  • Availability
  • Add to Cart