ENA390347533 Enamine

1-(3,3-dimethylpiperazin-1-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD19285713
  • Cas Number : 1268009-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126546

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126546
Availability details for
Quantity
  • Availability
  • Add to Cart