ENA390347534 Enamine

5-chloro-8-methylquinolin-2-amine

  • MDL number MFCD19678594
  • Cas Number : 1339793-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126547

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126547
Availability details for
Quantity
  • Availability
  • Add to Cart