ENA390347540 Enamine

2-[(2-bromophenyl)amino]cyclohexan-1-ol

  • MDL number MFCD12801881
  • Cas Number : 1183650-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126558
Availability details for
Quantity
  • Availability
  • Add to Cart