ENA390347545 Enamine

1-(4-methoxyphenyl)-4-(oxiran-2-ylmethyl)piperazine

  • MDL number MFCD09810238
  • Cas Number : 1016683-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126568
Availability details for
Quantity
  • Availability
  • Add to Cart