ENA390347549 Enamine

(4-chlorophenyl)[4-(trifluoromethyl)phenyl]methanone

  • MDL number MFCD00156808
  • Cas Number : 34328-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125132
Availability details for
Quantity
  • Availability
  • Add to Cart