ENA390347550 Enamine

1-(5-bromo-7-methyl-1-benzofuran-2-yl)methanamine

  • MDL number MFCD18294038
  • Cas Number : 1343809-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125133
Availability details for
Quantity
  • Availability
  • Add to Cart