ENA390347555 Enamine

4-chloro-2-(piperidin-1-yl)pyrimidine

  • MDL number MFCD00234559
  • Cas Number : 24192-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126600
Availability details for
Quantity
  • Availability
  • Add to Cart