ENA390347569 Enamine

1,3-bis(propan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD14609307
  • Cas Number : 72615-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126631

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126631
Availability details for
Quantity
  • Availability
  • Add to Cart