ENA390347571 Enamine

7,8-dichloroquinolin-2-amine

  • MDL number MFCD19678414
  • Cas Number : 1339883-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126636
Availability details for
Quantity
  • Availability
  • Add to Cart