ENA390347583 Enamine

4-(dimethylcarbamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08445050
  • Cas Number : 190711-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29868

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29868
Availability details for
Quantity
  • Availability
  • Add to Cart