ENA390347590 Enamine

2-chloro-N-(2-methoxyethyl)-N-(pentan-3-yl)acetamide

  • MDL number MFCD12762692
  • Cas Number : 1183841-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48532
Availability details for
Quantity
  • Availability
  • Add to Cart