ENA390347592 Enamine

2-[5-(chlorosulfonyl)-2-methoxyphenyl]acetic acid

  • MDL number MFCD09049843
  • Cas Number : 926244-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49333
Availability details for
Quantity
  • Availability
  • Add to Cart