ENA390347596 Enamine

1-(1H-pyrazol-1-yl)propan-2-one

  • MDL number MFCD03990541
  • Cas Number : 64882-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...182
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51213
Availability details for
Quantity
  • Availability
  • Add to Cart