ENA390347603 Enamine

1-phenyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD14657998
  • Cas Number : 60729-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59811
Availability details for
Quantity
  • Availability
  • Add to Cart