ENA390347614 Enamine

bis(piperidine-1-carboximidamide); sulfuric acid

  • MDL number MFCD01748702
  • Cas Number : 17238-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290-295
Purity 95
Salt Data 2H2SO4
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75399
Availability details for
Quantity
  • Availability
  • Add to Cart