ENA390347621 Enamine

2-[4-(hydroxymethyl)phenoxy]acetonitrile

  • MDL number MFCD11149454
  • Cas Number : 123226-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83408
Availability details for
Quantity
  • Availability
  • Add to Cart