ENA390347639 Enamine

2-(bromomethyl)but-1-ene

  • MDL number MFCD20621327
  • Cas Number : 59032-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95135

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-95135
pictograms GHS02,GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart