ENA390347644 Enamine

(oxolan-3-yl)(phenyl)methanamine

  • MDL number MFCD14610364
  • Cas Number : 1248371-63-2
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125154
Availability details for
Quantity
  • Availability
  • Add to Cart