ENA390347646 Enamine

4,6-dimethylpyridine-2-carbonitrile

  • MDL number MFCD08062830

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43...45
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97468
Availability details for
Quantity
  • Availability
  • Add to Cart