ENA390552127 Enamine

tert-butyl 2-(4,5-dihydrofuran-3-yl)-2-oxoacetate

  • MDL number MFCD24369082
  • Cas Number : 1432680-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117023
Availability details for
Quantity
  • Availability
  • Add to Cart