ENA390552130 Enamine

tert-butyl N-[6-(trifluoromethyl)piperidin-3-yl]carbamate

  • MDL number MFCD24279953
  • Cas Number : 1432681-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120865

Safety Information

signalword Warning
pcodes P261,P264,P271,P273,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-120865
pictograms GHS07,GHS09
hstatements H315,H319,H335,H400
Availability details for
Quantity
  • Availability
  • Add to Cart