ENA390552132 Enamine

5-cyclopropyl-1-propyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD24369088
  • Cas Number : 1432682-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123445
Availability details for
Quantity
  • Availability
  • Add to Cart