ENA390552133 Enamine

2-chloroprop-2-ene-1-sulfonamide

  • MDL number MFCD24369090
  • Cas Number : 1432682-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124019
Availability details for
Quantity
  • Availability
  • Add to Cart