ENA390552138 Enamine

1-(1-cyclopropylethyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD24369096
  • Cas Number : 1432680-82-4
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-cyclopropylethyl)-5-methyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C10H14N2O2/c1-6-5-9(10(13)14)11-12(6)7(2)8-3-4-8/h5,7-8H,3-4H2,1-2H3,(H,13,14)
Inchi Key DHYDNNBBTZBCNI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124358

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124358
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart