ENA390552140 Enamine

2-(pyrrolidin-2-yl)acetamide hydrochloride

  • MDL number MFCD24369098
  • Cas Number : 1432681-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124652
Availability details for
Quantity
  • Availability
  • Add to Cart