ENA390552143 Enamine

1-tert-butyl-3-methyl-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD24369101
  • Cas Number : 58046-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124919

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124919
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart