ENA390552144 Enamine

1-tert-butyl-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD24369102
  • Cas Number : 1432682-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124920

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124920
Availability details for
Quantity
  • Availability
  • Add to Cart