ENA390552146 Enamine

methyl 1-[4-(chlorosulfonyl)phenyl]cyclopropane-1-carboxylate

  • MDL number MFCD24369104
  • Cas Number : 600134-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125458

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125458
Availability details for
Quantity
  • Availability
  • Add to Cart