ENA390552147 Enamine

(4-chloro-2-cyanophenyl)methanesulfonyl chloride

  • MDL number MFCD24369105
  • Cas Number : 1432681-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125789

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-125789
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart