ENA390552148 Enamine

tert-butyl 4-(4-hydroxyphenyl)-1,2,3,6-tetrahydropyridine-1-carboxylate

  • MDL number MFCD24369106
  • Cas Number : 742067-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125815

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125815
Availability details for
Quantity
  • Availability
  • Add to Cart