ENA390552167 Enamine

3-chloro-4-fluoro-N-methylaniline hydrochloride

  • MDL number MFCD24369140
  • Cas Number : 1432680-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126888

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126888
Availability details for
Quantity
  • Availability
  • Add to Cart