ENA390552168 Enamine

2-(4-fluoro-3-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD24369141
  • Cas Number : 1375068-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126889
Availability details for
Quantity
  • Availability
  • Add to Cart