ENA390552171 Enamine

2-(propan-2-yl)morpholine hydrochloride

  • MDL number MFCD24369144
  • Cas Number : 1432678-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126932
Availability details for
Quantity
  • Availability
  • Add to Cart