ENA390552176 Enamine

3-fluoro-4-[2-(propan-2-yl)pyrrolidin-1-yl]aniline dihydrochloride

  • MDL number MFCD24369148
  • Cas Number : 1432679-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126959
Availability details for
Quantity
  • Availability
  • Add to Cart