ENA390552195 Enamine

4-(2,2-difluoroethoxy)pyridine-2-carboxylic acid

  • MDL number MFCD24253772
  • Cas Number : 1432681-88-3
  • Molecular Weight : 203.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2,2-difluoroethoxy)picolinic acid
Inchi Code 1S/C8H7F2NO3/c9-7(10)4-14-5-1-2-11-6(3-5)8(12)13/h1-3,7H,4H2,(H,12,13)
Inchi Key LBKTWMITDYKATQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127239
Availability details for
Quantity
  • Availability
  • Add to Cart