ENA390552197 Enamine

1,4-dioxaspiro[4.6]undecan-2-ylmethanamine

  • MDL number MFCD24304445
  • Cas Number : 46145-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127242
Availability details for
Quantity
  • Availability
  • Add to Cart