ENA390552199 Enamine

3-(2-ethoxyethoxy)azetidine hydrochloride

  • MDL number MFCD24369180
  • Cas Number : 1432681-92-9
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-ethoxyethoxy)azetidine hydrochloride
Inchi Code 1S/C7H15NO2.ClH/c1-2-9-3-4-10-7-5-8-6-7;/h7-8H,2-6H2,1H3;1H
Inchi Key SVNIICQIEAAEGW-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127247

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-127247
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart