ENA390552205 Enamine

4-hydrazinyl-1lambda6-thiane-1,1-dione hydrochloride

  • MDL number MFCD24369186
  • Cas Number : 1432678-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127301
Availability details for
Quantity
  • Availability
  • Add to Cart