ENA390552212 Enamine

1-[3-(trifluoromethyl)phenyl]prop-2-en-1-amine hydrochloride

  • MDL number MFCD24369194
  • Cas Number : 1432679-86-1
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127352
Availability details for
Quantity
  • Availability
  • Add to Cart