ENA390552215 Enamine

2-(3,4-dichlorophenyl)-2,2-difluoroacetonitrile

  • MDL number MFCD24369197
  • Cas Number : 1432681-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127397
Availability details for
Quantity
  • Availability
  • Add to Cart