ENA390552216 Enamine

[4-(2,2-difluoroethoxy)pyridin-2-yl]methanamine

  • MDL number MFCD24253771
  • Cas Number : 1432680-47-1
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-(2,2-difluoroethoxy)pyridin-2-yl)methanamine
Inchi Code 1S/C8H10F2N2O/c9-8(10)5-13-7-1-2-12-6(3-7)4-11/h1-3,8H,4-5,11H2
Inchi Key OEJSCBJWSVWOTD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127401
Availability details for
Quantity
  • Availability
  • Add to Cart